Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFESD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001131065.1 | NP_001124537.1 | 6 | 2195 | ||
NM_001131066.1 | NP_001124538.1 | 6 | 1040 | |||
NM_173362.3 | NP_775498.1 | 5 | 2097 | |||
XM_011543360.2 | XP_011541662.1 | 7 | 2083 | |||
XM_011543361.2 | XP_011541663.1 | 5 | 975 | |||
XM_017009413.1 | XP_016864902.1 | 6 | 1207 | |||
GenBank mRNA | BC024023.1 | - | 5 | 785 | 4670524 | |
BC035110.2 | - | 5 | 2095 | 5262922 |