Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MROH2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173489.4 | NP_775760.3 | 16 | 2139 | ||
XM_011513952.1 | XP_011512254.1 | 16 | 2125 | |||
XM_011513953.1 | XP_011512255.1 | 15 | 2063 | |||
GenBank mRNA | AK128183.1 | - | 6 | 605 | ||
AL832763.1 | - | 11 | 1064 | |||
BC044937.1 | - | 4 | 611 | |||
BC052962.1 | - | 16 | 2100 | 4838071 | ||
CR749346.1 | - | 11 | 1040 |