Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYCP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282541.1 | NP_001269470.1 | 4, 5 | 488 | ||
NM_001282542.1 | NP_001269471.1 | 4, 5 | 488 | |||
NM_003176.3 | NP_003167.2 | 4, 5 | 485 | |||
NR_104211.1 | 4, 5 | 485 | ||||
XM_005271156.1 | XP_005271213.1 | 4, 5 | 448 | |||
XM_006710859.1 | XP_006710922.1 | 4, 5 | 396 | |||
XM_011542037.2 | XP_011540339.1 | 4, 5 | 452 | |||
XM_011542039.1 | XP_011540341.1 | 4, 5 | 452 | |||
XM_017002184.1 | XP_016857673.1 | 4, 5 | 288 | |||
GenBank mRNA | AF273043.1 | - | 4, 5 | 423 | ||
AK315268.1 | - | 4, 5 | 447 | |||
BC105299.1 | - | 4, 5 | 378 | |||
BC126266.1 | - | 4, 5 | 362 | 8991982 | ||
BC144019.1 | - | 4, 5 | 365 | 9052539 | ||
BG772129.1 | - | 4, 5 | 460 | |||
BI561137.1 | - | 4, 5 | 477 | |||
D67035.1 | - | 4, 5 | 421 | |||
X95654.1 | - | 4, 5 | 339 |