Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM153C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC101338.1 | - | 9 | 794 | 40025116 | |
BC101339.2 | - | 12 | 931 | |||
BC101340.2 | - | 10 | 706 | |||
BC101341.2 | - | 9 | 794 | 40025123 | ||
BC131737.1 | - | 10 | 706 | 40025113 | ||
BC146883.1 | - | 9 | 1006 | 9056811 | ||
BC146888.1 | - | 9 | 1006 | 9056816 | ||
BC171810.1 | - | 10 | 919 | 9054464 |
Gene Symbol: FAM153A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173663.3 | NP_775934.3 | 20 | 1075 | ||
GenBank mRNA | AF155097.1 | - | 20 | 1247 |
Gene Symbol: FAM153B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001265615.1 | NP_001252544.1 | 20 | 1075 |