Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM153C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC101338.1 | - | 9 | 1027 | 40025116 | |
BC101339.2 | - | 12 | 1164 | |||
BC101340.2 | - | 10 | 939 | |||
BC101341.2 | - | 9 | 1027 | 40025123 | ||
BC131737.1 | - | 10 | 939 | 40025113 |
Gene Symbol: FAM153A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173663.3 | NP_775934.3 | 20 | 1310 | ||
GenBank mRNA | AF155097.1 | - | 20 | 1482 |
Gene Symbol: FAM153B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001265615.1 | NP_001252544.1 | 20 | 1310 |