Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTWD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308081.1 | NP_001295010.1 | 6 | 971 | ||
NM_173666.3 | NP_775937.1 | 6 | 1192 | |||
XM_011543338.2 | XP_011541640.2 | 7 | 2585 | |||
XM_011543340.2 | XP_011541642.1 | 7 | 1084 | |||
GenBank mRNA | AK091296.1 | - | 6 | 1135 | ||
BC114508.1 | - | 6 | 1128 | 40024023 | ||
BC114955.1 | - | 6 | 1128 | 40024022 | ||
BX648369.1 | - | 6 | 971 |