Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTWD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308081.1 | NP_001295010.1 | 6 | 1836 | ||
NM_173666.3 | NP_775937.1 | 6 | 2057 | |||
XM_011543338.2 | XP_011541640.2 | 7 | 3450 | |||
XM_011543340.2 | XP_011541642.1 | 7 | 1949 | |||
GenBank mRNA | AK091296.1 | - | 6 | 2000 | ||
BC012294.1 | - | 1 | 228 | |||
BX648369.1 | - | 6 | 1836 |