Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304548.1 | NP_001291477.1 | 31 | 5040 | ||
XM_017029452.1 | XP_016884941.1 | 31 | 5033 | |||
XM_017029453.1 | XP_016884942.1 | 31 | 5033 | |||
GenBank mRNA | AK093920.1 | - | 6 | 885 | ||
BC101698.1 | - | 5 | 739 | 8069204 | ||
BC101700.1 | - | 5 | 739 | 8069206 |