Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGEB18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173699.3 | NP_775970.1 | 3 | 1522 | ||
GenBank mRNA | AK057361.1 | - | 3 | 1469 | ||
BC029525.1 | - | 3 | 1522 | 5297993 |