Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCCHC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001312891.1 | NP_001299820.1 | 3 | 1530 | ||
NM_173798.3 | NP_776159.1 | 4 | 1582 | |||
GenBank mRNA | AK122676.1 | - | 4 | 1542 | ||
BC031241.1 | - | 4 | 1501 | 5263970 | ||
BC036572.2 | - | 4 | 1535 | 5274563 | ||
BC057841.1 | - | 4 | 1501 | 5264353 |