Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM84B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174911.4 | NP_777571.1 | 2 | 804 | ||
XM_017013107.1 | XP_016868596.1 | 2 | 851 | |||
XM_017013108.1 | XP_016868597.1 | 3 | 783 | |||
GenBank mRNA | AJ417849.1 | - | 2 | 731 | ||
AK292824.1 | - | 2 | 729 | |||
BC052957.1 | - | 2 | 559 | 5262890 |