Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOWAHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175873.5 | NP_787069.4 | 1 | 1975 | ||
GenBank mRNA | AK090823.1 | - | 1 | 402 | ||
AK092782.1 | - | 1 | 918 | |||
BC104771.1 | - | 1 | 1104 | 8143774 | ||
BC104775.1 | - | 1 | 1104 | 8143778 |