Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206855.1 | NP_001193784.1 | 3, 4 | 478 | ||
NM_003227.3 | NP_003218.2 | 6, 7 | 898 | |||
XM_005250553.4 | XP_005250610.1 | 7, 8 | 1204 | |||
XM_017012573.1 | XP_016868062.1 | 7, 8 | 1006 | |||
GenBank mRNA | AF067864.1 | - | 6, 7 | 884 | ||
AF085928.1 | - | 3, 4 | 341 | |||
AK000421.1 | - | 1, 2 | 112 | |||
BC142630.1 | - | 6, 7 | 884 | 40147690 | ||
BX384750.2 | - | 3, 4 | 478 |