Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC71L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175884.4 | NP_787080.2 | 1 | 1069 | ||
GenBank mRNA | AK093350.1 | - | 1 | 765 | ||
BC013906.2 | - | 2 | 772 | 3637017 |