Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRIPAK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175918.3 | NP_787114.2 | 1 | 4305 | ||
GenBank mRNA | AK091762.1 | - | 1 | 1812 | ||
AK096209.1 | - | 1 | 1493 | |||
AK126919.1 | - | 1 | 4310 | |||
AK128809.1 | - | 3504 | ||||
BC131709.1 | - | 2 | 1647 |