Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF800 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176814.4 | NP_789784.2 | 5 | 1706 | ||
XM_005250181.3 | XP_005250238.1 | 5 | 1780 | |||
XM_006715867.3 | XP_006715930.1 | 5 | 1785 | |||
XM_006715868.3 | XP_006715931.1 | 5 | 1780 | |||
XM_006715869.3 | XP_006715932.1 | 4 | 1689 | |||
XM_011515859.2 | XP_011514161.1 | 5 | 1501 | |||
GenBank mRNA | AF218032.1 | - | 5 | 1676 | ||
AK297946.1 | - | 3 | 894 | |||
AK309674.1 | - | 5 | 1700 | |||
BC037348.1 | - | 1 | 680 | |||
BC047641.1 | - | 5 | 1725 | |||
BC110623.1 | - | 5 | 1251 | 40036036 | ||
BC110624.1 | - | 5 | 1246 | 40036038 |