Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113561.2 | NP_001107033.1 | 4 | 723 | ||
NM_001323292.1 | NP_001310221.1 | 4 | 723 | |||
NM_178532.4 | NP_848627.1 | 4 | 723 | |||
XM_017009383.1 | XP_016864872.1 | 4 | 723 | |||
XM_017009384.1 | XP_016864873.1 | 3 | 957 | |||
XM_017009385.1 | XP_016864874.1 | 3 | 956 | |||
XM_017009386.1 | XP_016864875.1 | 4 | 723 | |||
XM_017009387.1 | XP_016864876.1 | 4 | 723 | |||
XM_017009388.1 | XP_016864877.1 | 4 | 682 | |||
XM_017009389.1 | XP_016864878.1 | 4 | 719 | |||
GenBank mRNA | AK090756.1 | - | 4 | 722 | ||
AL832580.2 | - | 4 | 682 | |||
BC101277.2 | - | 4 | 686 | 40024386 | ||
BC101278.2 | - | 4 | 686 | 40024390 | ||
BC101397.2 | - | 4 | 682 | 40024392 |