Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178556.3 | NP_848651.2 | 6 | 1571 | ||
XM_006714207.3 | XP_006714270.1 | 7 | 1452 | |||
XM_011531919.1 | XP_011530221.1 | 7 | 1423 | |||
GenBank mRNA | AK093499.1 | - | 6 | 1571 | ||
BC015684.2 | - | 3 | 916 | |||
BC113860.1 | - | 6 | 1524 | 40033174 | ||
BC114469.1 | - | 6 | 1524 | 40033173 |