Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQUB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282855.1 | NP_001269784.1 | 2 | 893 | ||
NM_001321293.1 | NP_001308222.1 | 2 | 905 | |||
NM_178827.4 | NP_849149.3 | 2 | 493 | |||
NR_104244.1 | 2 | 480 | ||||
NR_104245.1 | 2 | 493 | ||||
XM_005250161.3 | XP_005250218.1 | 3 | 608 | |||
XM_005250162.4 | XP_005250219.1 | 2 | 474 | |||
XM_011515833.2 | XP_011514135.1 | 3 | 421 | |||
XM_011515834.2 | XP_011514136.1 | 2 | 726 | |||
XM_017011771.1 | XP_016867260.1 | 3 | 407 | |||
XM_017011772.1 | XP_016867261.1 | 2 | 906 | |||
GenBank mRNA | AK093153.1 | - | 2 | 413 | ||
AK127736.1 | - | 2 | 906 | |||
BC026173.1 | - | 2 | 484 | 4838069 | ||
BC032838.2 | - | 2 | 418 | |||
BC036121.1 | - | 2 | 893 | 5296826 | ||
BC068607.1 | - | 2 | 418 | |||
BC091520.1 | - | 2 | 348 | 30379054 | ||
BC128181.1 | - | 2 | 392 | 40113310 | ||
BC128182.1 | - | 2 | 392 | 40113312 | ||
DA112303.1 | - | 2 | 494 |