Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003241.3 | NP_003232.2 | 4, 5 | 513 | ||
XM_011534042.1 | XP_011532344.1 | 5, 6 | 662 | |||
GenBank mRNA | AK130286.1 | - | 1 | 537 | ||
AK300523.1 | - | 4, 5 | 382 | |||
AK312936.1 | - | 4, 5 | 511 | |||
BC007003.1 | - | 4, 5 | 508 | 3950865 | ||
DA870110.1 | - | 4, 5 | 513 | |||
L34840.1 | - | 4, 5 | 484 | |||
U31905.1 | - | 4, 5 | 484 | |||
U79008.1 | - | 3, 4 | 377 | |||
Y09559.1 | - | 3, 4 | 377 |