Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUS4L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270419.1 | NP_001257348.1 | 4 | 439 | ||
NM_181581.2 | NP_853559.1 | 4 | 589 | |||
NR_073002.1 | 4 | 627 | ||||
NR_073003.1 | 4 | 589 | ||||
NR_073004.1 | 4 | 589 | ||||
NR_073005.1 | 4 | 439 | ||||
XM_005250125.4 | XP_005250182.1 | 3 | 451 | |||
XM_011515766.2 | XP_011514068.1 | 4 | 583 | |||
XM_011515767.2 | XP_011514069.1 | 3 | 333 | |||
XM_017011708.1 | XP_016867197.1 | 4 | 604 | |||
XM_017011709.1 | XP_016867198.1 | 4 | 584 | |||
XM_017011710.1 | XP_016867199.1 | 4 | 438 | |||
XM_017011711.1 | XP_016867200.1 | 3 | 366 | |||
XM_017011712.1 | XP_016867201.1 | 3 | 350 | |||
XM_017011713.1 | XP_016867202.1 | 3 | 420 | |||
GenBank mRNA | AK127802.1 | - | 1 | 3631 | ||
AK294095.1 | - | 4 | 588 | |||
AK297195.1 | - | 4 | 596 | |||
AL523141.3 | - | 4 | 559 | |||
BC111774.1 | - | 3 | 925 | 40033792 | ||
BG250736.1 | - | 415 | ||||
BX395100.2 | - | 4 | 398 | |||
CX787147.1 | - | 2 | 158 | |||
DA223301.1 | - | 4 | 411 | |||
U62767.1 | - | 4 | 387 |