Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUS4L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270419.1 | NP_001257348.1 | 6 | 626 | ||
NM_181581.2 | NP_853559.1 | 6 | 776 | |||
NR_073002.1 | 6 | 876 | ||||
NR_073003.1 | 5 | 658 | ||||
NR_073004.1 | 5 | 658 | ||||
NR_073005.1 | 5 | 508 | ||||
XM_005250125.4 | XP_005250182.1 | 5 | 638 | |||
XM_006715844.2 | XP_006715907.1 | 4 | 514 | |||
XM_006715845.3 | XP_006715908.1 | 3 | 425 | |||
XM_011515766.2 | XP_011514068.1 | 6 | 770 | |||
XM_011515767.2 | XP_011514069.1 | 5 | 520 | |||
XM_017011708.1 | XP_016867197.1 | 6 | 791 | |||
XM_017011709.1 | XP_016867198.1 | 6 | 771 | |||
XM_017011710.1 | XP_016867199.1 | 6 | 625 | |||
XM_017011711.1 | XP_016867200.1 | 5 | 553 | |||
XM_017011712.1 | XP_016867201.1 | 5 | 537 | |||
XM_017011713.1 | XP_016867202.1 | 5 | 607 | |||
GenBank mRNA | AK127802.1 | - | 2 | 3700 | ||
AK294095.1 | - | 5 | 657 | |||
AK297195.1 | - | 6 | 845 | |||
AK308472.1 | - | 1 | 352 | |||
AW954755.1 | - | 1 | 61 | |||
BC111774.1 | - | 4 | 994 | 40033792 | ||
BG250736.1 | - | 602 | ||||
CX787147.1 | - | 4 | 345 | |||
DA223301.1 | - | 5 | 480 | |||
U62767.1 | - | 6 | 574 |