Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUS4L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270419.1 | NP_001257348.1 | 8 | 1137 | ||
NM_181581.2 | NP_853559.1 | 8 | 1287 | |||
NR_073002.1 | 8 | 1387 | ||||
NR_073003.1 | 7 | 1169 | ||||
NR_073004.1 | 7 | 1126 | ||||
NR_073005.1 | 7 | 1019 | ||||
XM_005250125.4 | XP_005250182.1 | 7 | 1149 | |||
XM_006715844.2 | XP_006715907.1 | 6 | 1025 | |||
XM_006715845.3 | XP_006715908.1 | 5 | 936 | |||
XM_011515766.2 | XP_011514068.1 | 8 | 1281 | |||
XM_011515767.2 | XP_011514069.1 | 7 | 1031 | |||
XM_017011708.1 | XP_016867197.1 | 8 | 1302 | |||
XM_017011709.1 | XP_016867198.1 | 8 | 1282 | |||
XM_017011710.1 | XP_016867199.1 | 8 | 1136 | |||
XM_017011711.1 | XP_016867200.1 | 7 | 1064 | |||
XM_017011712.1 | XP_016867201.1 | 7 | 1048 | |||
XM_017011713.1 | XP_016867202.1 | 7 | 1118 | |||
GenBank mRNA | AK294095.1 | - | 7 | 1125 | ||
AK297195.1 | - | 8 | 1356 | |||
AK308472.1 | - | 3 | 863 | |||
BC111774.1 | - | 6 | 1505 | 40033792 | ||
BX357552.2 | - | 4 | 602 | |||
U62767.1 | - | 8 | 1085 |