Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYRM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181705.3 | NP_859056.2 | 4, 5 | 346 | ||
GenBank mRNA | AK000803.1 | - | 4, 5 | 317 | ||
BC037930.1 | - | 4, 5 | 298 | 5284728 | ||
BC047079.1 | - | 4, 5 | 276 | 5286889 | ||
BI601105.1 | - | 4, 5 | 322 | |||
DA996312.1 | - | 4, 5 | 348 |