Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGEB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182506.3 | NP_872312.2 | 3 | 769 | ||
GenBank mRNA | AK057527.1 | - | 3 | 769 | ||
BC101307.2 | - | 3 | 751 | 40024749 | ||
BC101308.2 | - | 3 | 751 | 40024751 | ||
BC101309.3 | - | 3 | 751 | 40024752 | ||
BC101310.2 | - | 3 | 751 | 40024754 |