Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003254.2 | NP_003245.1 | 5 | 539 | ||
XM_017029766.1 | XP_016885255.1 | 4 | 704 | |||
GenBank mRNA | A10416.1 | - | 5 | 409 | ||
AK074854.1 | - | 5 | 517 | |||
AK296116.1 | - | 3 | 893 | |||
AK310206.1 | - | 4 | 602 | |||
AK311937.1 | - | 5 | 391 | |||
BC000866.1 | - | 5 | 492 | 3460153 | ||
BC007097.1 | - | 5 | 440 | |||
BU857950.1 | - | 586 | ||||
CR407638.1 | - | 4 | 347 | |||
CR541982.1 | - | 4 | 347 | |||
M12670.1 | - | 5 | 395 | |||
M59906.1 | - | 5 | 390 | |||
S68252.1 | - | 4 | 350 | |||
X02598.1 | - | 5 | 388 | |||
X03124.1 | - | 5 | 409 |