Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194291.2 | NP_919267.2 | 6 | 1132 | ||
XM_011516847.2 | XP_011515149.1 | 6 | 1126 | |||
XM_017013104.1 | XP_016868593.1 | 4 | 192 | |||
GenBank mRNA | BC017881.1 | - | 4 | 236 | ||
BC032396.1 | - | 6 | 433 | 5197382 | ||
BC041379.1 | - | 6 | 1129 | 5275048 |