Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYCP2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040274.2 | NP_001035364.2 | 13 | 1232 | ||
GenBank mRNA | AK128130.1 | - | 12 | 1186 | ||
BC012225.1 | - | 3 | 129 | |||
DA620756.1 | - | 4 | 206 |