Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF212 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001131034.3 | NP_001124506.1 | 10 | 2044 | ||
NM_194439.4 | NP_919420.1 | 10 | 1991 | |||
XM_011513439.1 | XP_011511741.1 | 11 | 2233 | |||
XM_011513443.1 | XP_011511745.1 | 9 | 2212 | |||
XM_011513445.2 | XP_011511747.1 | 5 | 1792 | |||
XM_017008039.1 | XP_016863528.1 | 10 | 2070 | |||
XM_017008040.1 | XP_016863529.1 | 11 | 2108 | |||
GenBank mRNA | AK096160.1 | - | 10 | 1990 | ||
BC050356.1 | - | 10 | 2002 | 4815093 |