Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXorf23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198279.3 | NP_938020.2 | 9 | 1844 | ||
XM_005274473.3 | XP_005274530.1 | 10 | 2488 | |||
XM_005274474.3 | XP_005274531.1 | 10 | 2428 | |||
XM_011545475.2 | XP_011543777.1 | 10 | 2098 | |||
XM_011545476.2 | XP_011543778.1 | 10 | 2065 | |||
XM_011545477.2 | XP_011543779.1 | 10 | 2399 | |||
XM_011545478.2 | XP_011543780.1 | 10 | 2488 | |||
XM_017029390.1 | XP_016884879.1 | 10 | 2339 | |||
GenBank mRNA | AK127709.1 | - | 8 | 1547 | ||
AL833278.1 | - | 8 | 1694 | |||
BC126172.1 | - | 7 | 979 |