Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXorf23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198279.3 | NP_938020.2 | 11 | 2049 | ||
XM_005274473.3 | XP_005274530.1 | 12 | 2693 | |||
XM_005274474.3 | XP_005274531.1 | 12 | 2633 | |||
XM_005274475.3 | XP_005274532.1 | 11 | 2603 | |||
XM_011545475.2 | XP_011543777.1 | 12 | 2303 | |||
XM_011545476.2 | XP_011543778.1 | 12 | 2270 | |||
XM_011545477.2 | XP_011543779.1 | 12 | 2604 | |||
XM_011545478.2 | XP_011543780.1 | 11 | 2537 | |||
XM_011545479.2 | XP_011543781.1 | 11 | 2515 | |||
XM_011545482.2 | XP_011543784.1 | 10 | 2358 | |||
XM_017029390.1 | XP_016884879.1 | 12 | 2544 | |||
XM_017029391.1 | XP_016884880.1 | 11 | 2543 | |||
GenBank mRNA | AK127709.1 | - | 10 | 1752 | ||
AL833278.1 | - | 10 | 1899 | |||
BC126172.1 | - | 9 | 1184 | |||
BC143906.1 | - | 7 | 938 |