Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLRG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198508.2 | NP_940910.1 | 1 | 198 | ||
XM_005250311.3 | XP_005250368.1 | 1 | 203 | |||
XM_011516140.2 | XP_011514442.1 | 1 | 203 | |||
XM_011516141.2 | XP_011514443.1 | 1 | 203 | |||
GenBank mRNA | AK126174.1 | - | 1 | 203 | ||
BC104963.1 | - | 1 | 163 | 8143966 | ||
BC104965.1 | - | 1 | 163 | 8143968 | ||
BC110858.1 | - | 1 | 150 | 6139360 | ||
DB128604.1 | - | 1 | 198 |