Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM7SF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277233.1 | NP_001264162.1 | 4, 5 | 669 | ||
NM_003273.3 | NP_003264.2 | 4, 5 | 669 | |||
NR_102367.1 | 3, 4 | 614 | ||||
GenBank mRNA | AF023676.1 | - | 2, 3 | 1163 | ||
AF096304.1 | - | 4, 5 | 633 | |||
AK290935.1 | - | 4, 5 | 647 | |||
AK296441.1 | - | 3, 4 | 590 | |||
BC009052.1 | - | 4, 5 | 635 | 3874367 | ||
BC012857.1 | - | 4, 5 | 597 | 4214662 | ||
BC038353.1 | - | 4, 5 | 639 | 3861907 | ||
BG771219.1 | - | 4, 5 | 655 |