Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC172 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198515.2 | NP_940917.1 | 9 | 1330 | ||
XM_006717820.3 | XP_006717883.1 | 9 | 1625 | |||
XM_017016206.1 | XP_016871695.1 | 9 | 1248 | |||
GenBank mRNA | AJ457798.1 | - | 8 | 1312 | ||
BC044830.1 | - | 9 | 1330 | 5166167 |