Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM173B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258388.1 | NP_001245317.1 | 4 | 527 | ||
NM_001258389.1 | NP_001245318.1 | 5 | 534 | |||
NM_199133.3 | NP_954584.2 | 5 | 578 | |||
NR_047668.1 | 4 | 552 | ||||
NR_047669.1 | 6 | 781 | ||||
NR_047670.1 | 6 | 670 | ||||
XM_011513964.1 | XP_011512266.1 | 5 | 581 | |||
GenBank mRNA | AK055604.1 | - | 6 | 658 | ||
AK123462.1 | - | 1 | 613 | |||
AK300042.1 | - | 5 | 555 | |||
AK302012.1 | - | 4 | 516 | |||
AK316317.1 | - | 5 | 563 | |||
BC063406.1 | - | 5 | 546 | 5190223 | ||
DA523490.1 | - | 5 | 513 | |||
DA843869.1 | - | 4 | 528 |