Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM173B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258388.1 | NP_001245317.1 | 4 | 920 | ||
NM_001258389.1 | NP_001245318.1 | 5 | 927 | |||
NM_199133.3 | NP_954584.2 | 5 | 971 | |||
NR_047668.1 | 4 | 945 | ||||
NR_047669.1 | 6 | 1174 | ||||
NR_047670.1 | 6 | 1063 | ||||
XM_011513964.1 | XP_011512266.1 | 5 | 974 | |||
GenBank mRNA | AK055604.1 | - | 6 | 1051 | ||
AK123462.1 | - | 1 | 1006 | |||
AK300042.1 | - | 5 | 948 | |||
AK302012.1 | - | 4 | 909 | |||
BC063406.1 | - | 5 | 939 | 5190223 |