Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNNC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003279.2 | NP_003270.1 | 4, 5 | 388 | ||
XM_011529031.2 | XP_011527333.1 | 4, 5 | 398 | |||
GenBank mRNA | AK291323.1 | - | 4, 5 | 386 | ||
AK300665.1 | - | 386 | ||||
BC005323.1 | - | 4, 5 | 386 | 3934188 | ||
BU854061.1 | - | 4, 5 | 419 | |||
CR541864.1 | - | 3, 4 | 322 | |||
CR541884.1 | - | 3, 4 | 322 | |||
M22307.1 | - | 4, 5 | 369 | |||
X07898.1 | - | 4, 5 | 386 |