Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079821.2 | NP_001073289.1 | 11 | 3528 | ||
NM_001127461.2 | NP_001120933.1 | 9 | 3999 | |||
NM_001127462.2 | NP_001120934.1 | 8 | 3965 | |||
NM_001243133.1 | NP_001230062.1 | 9 | 4136 | |||
NM_004895.4 | NP_004886.3 | 9 | 4136 | |||
NM_183395.2 | NP_899632.1 | 7 | 3794 | |||
XM_011544048.2 | XP_011542350.1 | 10 | 4275 | |||
XM_011544053.2 | XP_011542355.1 | 8 | 3821 | |||
XM_011544055.2 | XP_011542357.1 | 7 | 3650 | |||
XM_017000181.1 | XP_016855670.1 | 11 | 3752 | |||
XM_017000182.1 | XP_016855671.1 | 11 | 3697 | |||
XM_017000183.1 | XP_016855672.1 | 9 | 3992 | |||
XM_017000184.1 | XP_016855673.1 | 8 | 3821 | |||
GenBank mRNA | AB208891.1 | - | 8 | 3974 | ||
AF054176.1 | - | 5 | 1869 | |||
AF410477.1 | - | 9 | 4136 | |||
AF420469.1 | - | 10 | 3522 | |||
AF427617.1 | - | 7 | 3794 | |||
AF468522.1 | - | 9 | 3552 | |||
AY092033.1 | - | 7 | 3210 |