Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSX2IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166293.1 | NP_001159765.1 | 4 | 691 | ||
NM_001166294.1 | NP_001159766.1 | 3 | 559 | |||
NM_001166295.1 | NP_001159767.1 | 3 | 687 | |||
NM_001166417.1 | NP_001159889.1 | 5 | 869 | |||
NM_014021.3 | NP_054740.3 | 5 | 741 | |||
XM_005270427.1 | XP_005270484.1 | 4 | 630 | |||
XM_005270428.1 | XP_005270485.1 | 4 | 588 | |||
XM_005270429.1 | XP_005270486.1 | 4 | 630 | |||
XM_006710331.1 | XP_006710394.1 | 4 | 548 | |||
XM_011540610.1 | XP_011538912.1 | 5 | 638 | |||
XM_011540612.2 | XP_011538914.1 | 5 | 679 | |||
XM_017000234.1 | XP_016855723.1 | 4 | 619 | |||
XM_017000235.1 | XP_016855724.1 | 4 | 597 | |||
XM_017000236.1 | XP_016855725.1 | 4 | 613 | |||
XM_017000237.1 | XP_016855726.1 | 3 | 402 | |||
GenBank mRNA | AB023140.1 | - | 4 | 679 | ||
AK021515.1 | - | 2 | 1263 | |||
AK093173.1 | - | 5 | 626 | |||
AK292475.1 | - | 4 | 579 | |||
AK294057.1 | - | 3 | 446 | |||
AK316472.1 | - | 3 | 465 | |||
AL133046.1 | - | 5 | 681 | |||
AY367055.1 | - | 5 | 604 | |||
AY651262.1 | - | 2 | 264 | |||
BC033637.1 | - | 4 | 566 | 5578582 | ||
BC035580.1 | - | 3 | 500 | |||
BC064389.1 | - | 4 | 655 | 5745142 |