Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASXL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015338.5 | NP_056153.2 | 5 | 769 | ||
XM_006723727.3 | XP_006723790.1 | 4 | 1048 | |||
XM_006723728.3 | XP_006723791.1 | 5 | 378 | |||
XM_006723730.3 | XP_006723793.1 | 5 | 574 | |||
XM_006723732.2 | XP_006723795.1 | 5 | 550 | |||
XM_011528648.2 | XP_011526950.1 | 5 | 693 | |||
XM_011528652.2 | XP_011526954.1 | 6 | 1711 | |||
XM_017027704.1 | XP_016883193.1 | 5 | 1834 | |||
XM_017027705.1 | XP_016883194.1 | 5 | 847 | |||
XM_017027706.1 | XP_016883195.1 | 5 | 1051 | |||
GenBank mRNA | AB084280.1 | - | 4 | 322 | ||
AJ438952.2 | - | 5 | 595 | |||
AK122923.1 | - | 5 | 550 | |||
BC064984.1 | - | 5 | 848 | |||
BC137278.1 | - | 6 | 532 | 9021275 | ||
BC137280.1 | - | 6 | 532 | 9021277 |