Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASXL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015338.5 | NP_056153.2 | 10 | 1354 | ||
XM_006723727.3 | XP_006723790.1 | 9 | 1633 | |||
XM_006723728.3 | XP_006723791.1 | 10 | 963 | |||
XM_006723730.3 | XP_006723793.1 | 10 | 1159 | |||
XM_006723732.2 | XP_006723795.1 | 9 | 982 | |||
XM_006723733.1 | XP_006723796.1 | 3 | 988 | |||
XM_011528648.2 | XP_011526950.1 | 10 | 1278 | |||
XM_011528652.2 | XP_011526954.1 | 11 | 2296 | |||
XM_017027704.1 | XP_016883193.1 | 10 | 2419 | |||
XM_017027705.1 | XP_016883194.1 | 10 | 1432 | |||
XM_017027706.1 | XP_016883195.1 | 9 | 1483 | |||
GenBank mRNA | AB023195.2 | - | 4 | 404 | ||
AB084280.1 | - | 9 | 907 | |||
AJ438952.2 | - | 10 | 1180 | |||
AK122923.1 | - | 9 | 982 | |||
AL117518.1 | - | 3 | 324 | |||
BC137278.1 | - | 11 | 1117 | 9021275 | ||
BC137280.1 | - | 11 | 1117 | 9021277 |