Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288564.1 | NP_001275493.1 | 6 | 650 | ||
NM_001288565.1 | NP_001275494.1 | 5 | 1153 | |||
NM_001288566.1 | NP_001275495.1 | 6 | 751 | |||
NM_001288567.1 | NP_001275496.1 | 6 | 965 | |||
NM_001288568.1 | NP_001275497.1 | 6 | 803 | |||
NM_001288569.1 | NP_001275498.1 | 6 | 664 | |||
NM_001288570.1 | NP_001275499.1 | 6 | 677 | |||
NM_001288571.1 | NP_001275500.1 | 6 | 743 | |||
NM_016456.4 | NP_057540.1 | 6 | 668 | |||
NR_109920.1 | 7 | 895 | ||||
NR_109921.1 | 5 | 1122 | ||||
XM_011509385.1 | XP_011507687.1 | 6 | 727 | |||
XM_011509386.2 | XP_011507688.1 | 6 | 702 | |||
XM_011509387.2 | XP_011507689.1 | 6 | 590 | |||
GenBank mRNA | AB013909.1 | - | 5 | 521 | ||
AF151020.1 | - | 7 | 640 | |||
AK054883.1 | - | 6 | 623 | |||
AK075335.1 | - | 6 | 578 | |||
AK091726.1 | - | 5 | 1038 | |||
AK094588.1 | - | 2 | 1971 | |||
AK303843.1 | - | 6 | 689 | |||
AL136600.1 | - | 6 | 607 | |||
AY138587.1 | - | 6 | 579 | |||
AY359012.1 | - | 6 | 642 | |||
BC001106.2 | - | 6 | 592 | 3506442 | ||
BM928361.1 | - | 7 | 818 | |||
CR457162.1 | - | 5 | 521 | |||
CR533467.1 | - | 5 | 527 |