Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNTA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003098.2 | NP_003089.1 | 8 | 2215 | ||
XM_005260517.1 | XP_005260574.1 | 8 | 2173 | |||
XM_011529007.1 | XP_011527309.1 | 8 | 2208 | |||
XM_011529008.1 | XP_011527310.1 | 8 | 2205 | |||
GenBank mRNA | AK095942.1 | - | 8 | 1775 | ||
BC026215.2 | - | 8 | 2055 | 4413197 | ||
U40571.1 | - | 8 | 1980 |