Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206846.1 | NP_001193775.1 | 2, 3 | 708 | ||
NM_018136.4 | NP_060606.3 | 2, 3 | 708 | |||
GenBank mRNA | AF509326.1 | - | 2, 3 | 451 | ||
AK125107.1 | - | 2, 3 | 708 | |||
AK226178.1 | - | 2, 3 | 641 | |||
AY099892.1 | - | 2, 3 | 451 | |||
AY101201.1 | - | 1, 2 | 118 | |||
AY367065.1 | - | 2, 3 | 451 | |||
AY971955.1 | - | 2, 3 | 451 | |||
AY971956.1 | - | 2, 3 | 451 | |||
AY971957.1 | - | 2, 3 | 451 |