Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206846.1 | NP_001193775.1 | 9, 10 | 3025 | ||
NM_018136.4 | NP_060606.3 | 9, 10 | 3025 | |||
GenBank mRNA | AF509326.1 | - | 9, 10 | 2768 | ||
AK001380.1 | - | 5, 6 | 700 | |||
AK125107.1 | - | 9, 10 | 3025 | |||
AK226178.1 | - | 9, 10 | 2958 | |||
AY099891.1 | - | 7, 8 | 1550 | |||
AY099893.1 | - | 7, 8 | 1550 | |||
AY367065.1 | - | 9, 10 | 2768 | |||
AY971956.1 | - | 9, 10 | 2768 | |||
BC034607.1 | - | 5, 6 | 644 |