Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102594.1 | NP_001096064.1 | 9 | 1911 | ||
NM_001102595.1 | NP_001096065.1 | 8 | 1746 | |||
NM_001102596.1 | NP_001096066.1 | 6 | 1334 | |||
NM_020892.2 | NP_065943.2 | 10 | 2031 | |||
XM_005250131.1 | XP_005250188.1 | 10 | 1922 | |||
XM_005250132.1 | XP_005250189.1 | 8 | 1665 | |||
XM_011515772.1 | XP_011514074.1 | 10 | 2093 | |||
XM_011515773.2 | XP_011514075.1 | 10 | 2520 | |||
XM_011515774.1 | XP_011514076.1 | 10 | 1846 | |||
XM_011515775.1 | XP_011514077.1 | 9 | 1940 | |||
XM_017011723.1 | XP_016867212.1 | 9 | 1969 | |||
XM_017011724.1 | XP_016867213.1 | 9 | 2400 | |||
XM_017011725.1 | XP_016867214.1 | 9 | 1818 | |||
XM_017011726.1 | XP_016867215.1 | 8 | 1770 | |||
XM_017011727.1 | XP_016867216.1 | 7 | 1584 | |||
GenBank mRNA | AB040961.1 | - | 9 | 1833 | ||
AK023924.1 | - | 8 | 1687 | |||
AK026252.1 | - | 3 | 964 | |||
AK293226.1 | - | 8 | 1500 | |||
AY225124.1 | - | 7 | 1475 | |||
AY225125.1 | - | 6 | 1334 | |||
BC008856.2 | - | 8 | 1666 | 4107018 | ||
BC018555.1 | - | 10 | 1985 | 4341568 | ||
BC026059.2 | - | 8 | 1666 | 4099111 | ||
BC064535.1 | - | 4 | 583 | |||
BC093079.1 | - | 9 | 1799 | 30512572 | ||
DQ010329.1 | - | 10 | 1985 |