Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022781.4 | NP_073618.3 | 3 | 536 | ||
NM_194328.2 | NP_919309.1 | 3 | 703 | |||
NM_194329.2 | NP_919310.1 | 2 | 386 | |||
NM_194330.2 | NP_919311.1 | 3 | 434 | |||
NM_194332.2 | NP_919313.1 | 2 | 553 | |||
XM_005251366.3 | XP_005251423.1 | 3 | 540 | |||
XM_005251367.3 | XP_005251424.1 | 3 | 438 | |||
XM_005251368.3 | XP_005251425.1 | 3 | 552 | |||
XM_006716721.3 | XP_006716784.1 | 2 | 400 | |||
XM_011517712.2 | XP_011516014.1 | 4 | 476 | |||
XM_011517713.2 | XP_011516015.1 | 3 | 540 | |||
XM_017014295.1 | XP_016869784.1 | 3 | 567 | |||
GenBank mRNA | AF394047.1 | - | 3 | 658 | ||
AK024996.1 | - | 1 | 46 | |||
AK093480.1 | - | 2 | 504 | |||
AK097653.1 | - | 3 | 402 | |||
AL136817.1 | - | 3 | 454 | |||
AV690059.2 | - | 2 | 277 | |||
AX748155.1 | - | 2 | 504 | |||
BC033786.1 | - | 2 | 386 | 5166325 | ||
BI520161.1 | - | 3 | 432 | |||
DA189510.1 | - | 2 | 504 | |||
DA483270.1 | - | 2 | 284 |