Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022781.4 | NP_073618.3 | 6 | 961 | ||
NM_194328.2 | NP_919309.1 | 6 | 1128 | |||
NM_194329.2 | NP_919310.1 | 5 | 811 | |||
NM_194330.2 | NP_919311.1 | 6 | 859 | |||
NM_194332.2 | NP_919313.1 | 5 | 978 | |||
XM_005251366.3 | XP_005251423.1 | 6 | 965 | |||
XM_005251367.3 | XP_005251424.1 | 6 | 863 | |||
XM_005251368.3 | XP_005251425.1 | 6 | 977 | |||
XM_006716721.3 | XP_006716784.1 | 5 | 825 | |||
XM_011517712.2 | XP_011516014.1 | 7 | 901 | |||
XM_011517713.2 | XP_011516015.1 | 6 | 965 | |||
XM_017014294.1 | XP_016869783.1 | 5 | 882 | |||
XM_017014295.1 | XP_016869784.1 | 6 | 992 | |||
XM_017014296.1 | XP_016869785.1 | 5 | 562 | |||
XM_017014297.1 | XP_016869786.1 | 5 | 558 | |||
GenBank mRNA | AF394047.1 | - | 6 | 1083 | ||
AK024996.1 | - | 4 | 471 | |||
AK093480.1 | - | 6 | 1027 | |||
AK097653.1 | - | 6 | 1021 | |||
AL136817.1 | - | 6 | 879 | |||
AX748155.1 | - | 6 | 1027 | |||
BC033786.1 | - | 5 | 811 | 5166325 | ||
BI520161.1 | - | 6 | 866 | |||
DA483270.1 | - | 5 | 707 |