Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031739.2 | NP_001026909.1 | 7 | 1561 | ||
NM_001168530.1 | NP_001162002.1 | 7 | 1592 | |||
NM_001168531.1 | NP_001162003.1 | 8 | 1809 | |||
NM_024087.2 | NP_076992.1 | 7 | 1622 | |||
XM_005274446.1 | XP_005274503.1 | 8 | 1748 | |||
XM_011545458.1 | XP_011543760.1 | 8 | 1661 | |||
XM_011545460.1 | XP_011543762.1 | 8 | 1722 | |||
XM_017029283.1 | XP_016884772.1 | 7 | 1528 | |||
XM_017029284.1 | XP_016884773.1 | 7 | 1441 | |||
XM_017029285.1 | XP_016884774.1 | 7 | 1589 | |||
GenBank mRNA | AK000643.1 | - | 7 | 1512 | ||
AL080091.2 | - | 7 | 1338 | |||
BC001244.1 | - | 7 | 1472 | 3462727 | ||
BC013172.1 | - | 7 | 1511 | 3917449 |