Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319296.1 | NP_001306225.1 | 5 | 842 | ||
NM_001319297.1 | NP_001306226.1 | 4 | 702 | |||
NM_001319298.1 | NP_001306227.1 | 5 | 807 | |||
NM_001319299.1 | NP_001306228.1 | 5 | 803 | |||
NM_001319300.1 | NP_001306229.1 | 5 | 760 | |||
NM_001319301.1 | NP_001306230.1 | 5 | 756 | |||
NM_024095.4 | NP_077000.1 | 4 | 706 | |||
GenBank mRNA | AF464877.1 | - | 4 | 598 | ||
AK024908.1 | - | 4 | 607 | |||
AK125596.1 | - | 1 | 1453 | |||
AK289957.1 | - | 4 | 673 | |||
AK297013.1 | - | 5 | 512 | |||
BC001321.1 | - | 4 | 613 | 3456049 | ||
CR457336.1 | - | 3 | 536 |